Process wafer carriers, 200 mm
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A192-81M-0215 A192-80M-0215 A200-80MG-0215 A19280MLZ0215
216-1576EA 0 EUR
216-1576 216-1629 216-2375 216-2374
Process wafer carriers, 200 mm
Wafertransportsystemen Wafer dragers
PFA, natural.
  • Open side walls allow solutions to pass through evenly and quickly
  • Low and high profile
  • 'H' bar for equipment interface
  • Locating track notches at the centre of the 13th pocket
  • Pins and holes on the top rail (unless noted)
These moulded PFA process wafer carriers are designed for wet chemical processing applications.
Opgelet: Articles may not be available in all countries. Please contact your local Avantor office.