Cable ties, releasable, Allplastik-Blitzbinder®
Leverancier: WUERTEMBERGISCHE ALLPLASTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABV212ROT ABV218ROT
129-4812EA 6.5 EUR
129-4812 129-4813
Cable ties, releasable, Allplastik-Blitzbinder®
Spanbanden
The Allplastik-Blitzbinder® quick fasteners are made of PE and have a special dual closure design.
  • Provides maximum strength and reliable closing when combined with a round profile
  • Can be opened again at any time
  • Allows a hanging loop to be formed
  • Temperature resistance: −40 to +70 °C
Ideal for use as sack fastener, bag fastener, releasable cable tie, plant tie, general-purpose binder, quick fastener, fixing tie, bag tie, scaffolding tie, tarpaulin tie, cable tie for bundling cable harnesses.
Certificaten: FDA compliant