Hermetic jars, wide mouth
Leverancier: LAMAPLAST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2452-1 2450-1 2462-1 2458-1 2454-1
LAMA2452-1EA 669 EUR
LAMA2452-1 LAMA2450-1 LAMA2462-1 LAMA2458-1 LAMA2454-1
Hermetic jars, wide mouth
Potjes Potjes voor algemeen gebruik
PP, white.
  • Self-sealing screw cap
  • Cap must be ordered seperately
  • Good chemical resistance, good temperature resistance
Wide universal containers for creams, semi solid substances, powders and granulates.