Manual injection valve with position sensing switches
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5063-6502 0101-1231 0101-1232 0101-1253 5065-9922
HEWL5063-6502EA 1655 EUR
HEWL5063-6502 HEWL0101-1231 HEWL0101-1232 HEWL0101-1253 HEWL5065-9922
Manual injection valve with position sensing switches
Ventielen Injectieventielen
The stainless steel 7725i and PEEK 9725i manual injection valves are suitable for large sample volumes, high flow rates and preparative columns sized 10 to 100 mm in diameter.