ESD occupational shoes, clogs, Boston
Leverancier: LIEFERSERVICE UND LOGISTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
061378/36 061378/37 061378/39 061378/38 061378/40 061378/41 061378/42 061388/41 061388/40 061388/37 061360/42 061388/39 061368/39 061368/38 061368/37 061368/36 061360/48 061360/47 061360/46 061360/45 061368/41 061360/43 061368/42 061360/41 061360/40 061360/39 061360/44 061380/39 061388/36 061380/48 061380/47 061380/46 061380/45 061380/44 061380/43 061380/42 061368/40 061380/40 061388/38 061370/48 061370/47 061370/46 061370/45 061370/44 061370/43 061370/42 061370/41 061370/40 061370/39 061380/41
LIEF061378/36EA 109 EUR
LIEF061378/36 LIEF061378/37 LIEF061378/39 LIEF061378/38 LIEF061378/40 LIEF061378/41 LIEF061378/42 LIEF061388/41 LIEF061388/40 LIEF061388/37 LIEF061360/42 LIEF061388/39 LIEF061368/39 LIEF061368/38 LIEF061368/37 LIEF061368/36 LIEF061360/48 LIEF061360/47 LIEF061360/46 LIEF061360/45 LIEF061368/41 LIEF061360/43 LIEF061368/42 LIEF061360/41 LIEF061360/40 LIEF061360/39 LIEF061360/44 LIEF061380/39 LIEF061388/36 LIEF061380/48 LIEF061380/47 LIEF061380/46 LIEF061380/45 LIEF061380/44 LIEF061380/43 LIEF061380/42 LIEF061368/40 LIEF061380/40 LIEF061388/38 LIEF061370/48 LIEF061370/47 LIEF061370/46 LIEF061370/45 LIEF061370/44 LIEF061370/43 LIEF061370/42 LIEF061370/41 LIEF061370/40 LIEF061370/39 LIEF061380/41
ESD occupational shoes, clogs, Boston
Schoenen en laarzen
Anatomically shaped ESD clog made from leather upper (2,8 to 3 mm thick cowhide) and an EVA sole.
  • Cork/latex core with covered with strong suede lining
  • Deep integrated insole with lengthwise and crosswise arches
  • Deep heel cup
  • Shock-absorbent EVA sole
  • Adjustable leather rist strap
Colour: Black/white/blue
Certificaten: EN ISO 61340