Tubings, insulated
Leverancier: LAUDA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LZS001 LZS007 LZS009 LZS021 LZS002 LZS019 LZS020 LZS022 LZS011 LZS012 LZS023
LAUDLZS001EA 82.8 EUR
LAUDLZS001 LAUDLZS007 LAUDLZS009 615-1138 LAUDLZS002 LAUDLZS019 LAUDLZS020 LAUDLZS022 LAUDLZS011 LAUDLZS012 LAUDLZS023
Tubings, insulated
Slangen Transfer Tubing
This insulated tubing is used with thermostats.
*Silicone tubes should not used in conjunction with silicone oils.