Cartridge filters, ProcessGard® PP
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PP2H010E1 PP03010E1 PP1H010E1
ENTEPP2H010E1EA 0 EUR
ENTEPP2H010E1 514-0740 ENTEPP1H010E1
Cartridge filters, ProcessGard® PP
Filters Filterpatronen
These cartridge filters are suited for acids, bases, process water, solvents, dyes and lacquers in flat panel display and chemical manufacturing.
  • Broad chemical and thermal compatibility
  • For applications in wet etching, stripping, developer and cleaning process
  • Pre and final filtration for solvents, photoresist and all pigment chemicals
Membrane, supports, core and sleeves: PP
Maximum operating temperature: 80 °C
Maximum forward differential pressure: 4,0 bar at 20 °C
Maximum reverse differential pressure: 2,5 bar at 20 °C
Articles may not be available in all countries. Please contact your local Avantor office.