Cartridge filters, Fluorogard® FP
Leverancier: Entegris
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Fluorogard®
ENTECWES720E1EA 0 EUR
ENTECWES720E1 ENTECWEC710E1 ENTECWEG010E1 ENTECWEG010T1Y ENTECWEH030T1 ENTECWEC020T1 ENTECWEV010T1 ENTECWEG010V1 ENTECWEG030T1 ENTECWEH010E1 ENTECWEC010T1Y ENTECWEV020E1 ENTECWEG720E1 ENTECWEA010E1 ENTECWEGF10E1 ENTECWEV710E1 ENTECWES710E1 ENTECWEG020E1 ENTECWEA010T1Y ENTECWEH020T1 ENTECWEAF10E1 ENTECWEA010T1 ENTECWEG020V1 ENTECWEA020V1 ENTECWEA710E1 ENTECWEG710E1 ENTECWEA020E1 ENTECWEZ020T1 ENTECWEV010E1 ENTECWES010E1 ENTECWEG030E1 ENTECWEV720E1 ENTECWEZF10E1 ENTECWEH010T1 ENTECWEAF20E1 ENTECWEA020T1 ENTECWEV030T1 ENTECWEG710V1 ENTECWEV010V1 ENTECWESF20E1 ENTECWECF10E1 ENTECWES010T1 ENTECWEC010T1 ENTECWEG010T1 ENTECWEH030E1 ENTECWES020E1 ENTECWEZ010T1 ENTECWEH010T1Y ENTECWEAF30E1 ENTECWEVF10E1 ENTECWEV730E1 ENTECWEA010V1 ENTECWEA030T1 ENTECWEZ010E1 ENTECWEV020T1 ENTECWES020T1 ENTECWESF10E1 ENTECWEHF10E1 ENTECWEG020T1 ENTECWES710T1 ENTECWEV010T1Y ENTECWEC010E1 ENTECWEGF20E1 ENTECWES030E1
Cartridge filters, Fluorogard® FP
Filters Filterpatronen
These filters can be used in solvent based photochemicals without the need for prewetting steps.
  • Excellent chemical compatibility
  • For use in POU filtration and chemical delivery system filtration
Membrane: Hydrophobic PTFE; Supports, core, sleeve and endcaps: PP
Maximum operating temperature: 90 °C
Maximum forward/reverse differential pressure: 3,4 bar at 25 °C