Disposable filters, Optimizer® DEV and DI
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CBUZ3L0P1 CBUV0S0S1 CBUV6S0P1 CBUV8S0P1 CBUZ0S0S1 CHUV0S0P1 CHUV8S0P1 CHUZ0S0S1 CHUZ3L0P1 CHUZ8S0P1 CBUV0S0P1 CBUZ8S0P1 CBUG0S0F1 CBUG0S0P1 CBUG0S0S1 CBUG3L0P1 CBUG8L0P1 CBUG8S0P1 CBUV2L0P1 CBUV3L0P1 CBUV8L0P1 CBUZ0L0S1 CBUZ0S0F1 CBUZ0S0P1 CBUZ2L0P1 CBUZ3L0F1 CBUZ3L0X1 CBUZ6S0P1 CBUZ8L0P1 CHUV0L0P1 CHUV0S0F1 CHUV0S0S1 CHUV2L0P1 CHUV3L0P1 CHUV3L0S1 CHUV6S0P1 CHUZ0L0P1 CHUZ0S0P1 CHUZ2L0P1 CHUZ3L0S1 CHUZ3L0X1 CHUZ6S0P1 CHUY0S0P1M CHUY3L0P1M CHUY6S0P1M
ENTECBUZ3L0P1EA 0 EUR
ENTECBUZ3L0P1 ENTECBUV0S0S1 ENTECBUV6S0P1 ENTECBUV8S0P1 514-0734 ENTECHUV0S0P1 ENTECHUV8S0P1 ENTECHUZ0S0S1 ENTECHUZ3L0P1 514-0736 ENTECBUV0S0P1 514-0735 ENTECBUG0S0F1 ENTECBUG0S0P1 ENTECBUG0S0S1 ENTECBUG3L0P1 ENTECBUG8L0P1 ENTECBUG8S0P1 ENTECBUV2L0P1 ENTECBUV3L0P1 ENTECBUV8L0P1 ENTECBUZ0L0S1 ENTECBUZ0S0F1 ENTECBUZ0S0P1 ENTECBUZ2L0P1 ENTECBUZ3L0F1 ENTECBUZ3L0X1 ENTECBUZ6S0P1 ENTECBUZ8L0P1 ENTECHUV0L0P1 ENTECHUV0S0F1 ENTECHUV0S0S1 ENTECHUV2L0P1 ENTECHUV3L0P1 ENTECHUV3L0S1 ENTECHUV6S0P1 ENTECHUZ0L0P1 ENTECHUZ0S0P1 ENTECHUZ2L0P1 ENTECHUZ3L0S1 ENTECHUZ3L0X1 ENTECHUZ6S0P1 ENTECHUY0S0P1M ENTECHUY3L0P1M ENTECHUY6S0P1M
Disposable filters, Optimizer® DEV and DI
Filters Filterpatronen
These point-of-use filters meet small to mid-volume photochemical filtration needs for positive developer and DI water applications. Both Optimizer® DEV filters (for developer applications) and Optimizer® DI filters (for DI water applications) require no pre-wetting, so the filters function immediately after installation. The design also limits the handling of hazardous chemicals during the installation and promotes a healthier workplace. Do not use either filter with aromatic hydrocarbons.
  • Manufactured, tested, and packaged in a world-class, ISO 9001 Quality Systems Standard facility
  • O-ringless connections avoid potential sources of contamination
Membrane: Patented surface-modified hydrophilic ultra-high molecular weight polyethylene (UHMWPE); Supports, core and sleeve: HDPE; Disposable shell: PP
Maximum operating temperature: 60 °C
Maximum forward differential pressure: 2,8 bar at 25 °C
Maximum reverse differential pressure: 2,8 bar at 25 °C
Articles may not be available in all countries. Please contact your local Avantor office.