Syringe pumps, PHD 22/2000 advanced
Leverancier: HARVARD APPARATUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70-2002 71-2000 70-2003 70-2006 70-2009
HARV70-2002EA 0 EUR
HARV70-2002 HARV71-2000 HARV70-2003 HARV70-2006 HARV70-2009
Syringe pumps, PHD 22/2000 advanced
Pompen Spuitpompen
The PHD 22/2000 syringe pump series gives you the lowest flow rates, the highest accuracy, the smoothest flow, advanced programmability from the keypad. Ideal for applications in drug/nutritional delivery, electrospinning, microfluidics/nanofluidics and mass spec calibration.
  • High accuracy and precision
  • Multi-syringe racks available for large capacity reservoir
  • Smooth flow over a wide flow rate range
The standard 2-syringe rack: holds two syringes from 0,5 µl or 140 ml.
The 4×140 multi-rack: holds four 60 ml or 140 ml plastic syringes only.
The 6/10 multi-rack: will hold up to 10 syringes from 0,5 µl to 20 ml and up to 6 syringes from 30 to 60 ml.
The microliter syringe rack: independently holds four microliter syringes, from 0,5 µl to 10 ml, enabling syringes of different sizes to run simultaneously.
The push/pull syringe pump: holds four syringes, two in each direction, for syringe sizes 0,5 µl to 140 ml.