Wound plasters, Aluderm®-aluplast waterproof
Leverancier: SOEHNGEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1009155 1009157
SOEH1009155EA 0 EUR
SOEH1009155 SOEH1009157
Wound plasters, Aluderm®-aluplast waterproof
Eerste-hulpbenodigdheden Wondverbanden
The aluderm®-aluplast wound plaster has an aluminium wound layer and an additional absorption middle layer. The aluminised wound layer is highly absorbent, so the wound remains dry and clean.
  • Made from PE plastic with polyacryl adhesive
  • Adhesive function is activated by body heat
  • Plaster is transparent, durable and waterproof
  • Recommended in places where water and dirt are present
  • Individually sealed