Sterile Serological Pipettes
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0030127.692 0030127.706 0030127.714 0030127.722 0030127.730 0030127.749
EPPE0030127.692EA 163 EUR
EPPE0030127.692 EPPE0030127.706 EPPE0030127.714 EPPE0030127.722 EPPE0030127.730 EPPE0030127.749
Sterile Serological Pipettes
Pipetten Serologische pipetten
PS, transparant, steriel. Deze serologische pipetten zijn ontworpen voor gebruik in combinatie met de Easypet 3 controller. De pipetten bieden een nauwkeurige schaalverdeling op het uiterst heldere materiaal om het volume snel en eenvoudig af te lezen.
  • Clear and precise graduations for easy volume determination
  • Color-coded for easy identification of pipet sizes
  • Subunits with dispenser option to keep stored products safe
  • Individually wrapped for reliable sterility
  • Ultra-pure virgin polystyrene meets the requirement of USP VI
  • Harmonized and well-fitting operation with Easypet 3; compatibility with all existing pipet helpers
  • Sterility assurance level of 10⁻⁶
  • Certified free of detectable pyrogens, DNA, RNase, and DNase
  • Certified non-cytotoxic
Designed to work in perfect harmony with the Easypet 3 or Pipet Helper, Eppendorf completes its portfolio in liquid handling with this serological pipette. For ease and flexibility, these serological pipettes can also be used with any other standard pipette controller.
Rely on precise graduations overlaid on ultra-clear pipette material for quick and easy volume readings from 0,1 to 50 ml. Enjoy the freedom of individually wrapped pipettes packaged in a robust dispenser box to ensure consistent premium quality.