Volumetric flasks, class A, blue markings
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0293-04-0150 0293-04-035014
NEUB0293-04-0150EA 52.7 EUR
NEUB0293-04-0150 NEUB0293-04-035014
Volumetric flasks, class A, blue markings
Kolven Maatkolven
Borosilicate glass, grade 3.3, with PE stopper.
  • With one mark
  • Good chemical resistance
  • Good temperature resistance