In-line filter units, Millex®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SLFG85010 SLFG85000
514-0413EA 185 EUR
514-0413 MILFSLFG85000
In-line filter units, Millex®
Filters Lijnfilters
These filter units are ideal for sterilising gases, venting sterile containers and sterilising or clarifying organic solutions. Specialised filter units protect hemodialysis transducers from blood and moisture. 50 mm filter units are also used for vacuum line protection.
  • All units are bi-directional
PTFE hydrophobic Fluoropore membrane with PP housing
Maximum inlet pressure: 4,1 bar
Maximum operating temperature: 121 °C
Inlet/outlet connections: stepped hose barb with female Luer slip interior