Scoops, Fluo-Kem®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H367330000 H367350000
BELAH367330000EA 396 EUR
BELAH367330000 BELAH367350000
Scoops, Fluo-Kem®, SP Bel-Art
Scheppen
Aluminium, PTFE coated.
  • Uncoated handle allows non slip grip even with gloved hand
  • Complies with FDA regulations for food contact
  • PTFE coating resists corrosion and prevents samples from sticking