Burette, Sofnol, auto zero, MBL®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRB004 BRB006
BIBBBRB004EA 133 EUR
BIBBBRB004 BIBBBRB006
Burette, Sofnol, auto zero, MBL®
Buretten
Borosilicate glass, class B.
  • Blue enamel graduations
  • Easy filling and automatic zero
  • With simple rubber tube and pinch valve for low cost replacement
The burette can be filled by squeezing the bottle.
Complies with ISO 385