Dropping funnels, cylindrical, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D1/21 D1/22
532-0075EA 87.8 EUR
532-0075 532-0076
Dropping funnels, cylindrical, Quickfit®
Trechters Druppeltrechters
Funnel stem with ground-glass cone for flasks and assemblies. Stopcock has interchangeable glass key.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Certificaten: Complies with ISO 4800