Test tube racks
Leverancier: WDF - Funke GmbH & Co. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5270-312-13 5270-412-13 527-412-11 5273-26-11 527-1010-11 5270-1010-11 527-412-13 5278-412-13 527-34-13 527-26-11 527-46-11
212-6433EA 0 EUR
212-6433 212-6434 WCDF527-412-11 WCDF5273-26-11 WCDF527-1010-11 WCDF5270-1010-11 WCDF527-412-13 WCDF5278-412-13 WCDF527-34-13 WCDF527-26-11 WCDF527-46-11
Test tube racks
Rekken Tube Racks
Test tube racks in stainless steel, white PE, stainless steel or white PA.
  • PE white is heat resistant up to 60 °C
  • Stainless steel is electrolytically polished
  • PA white is permanently heat resistant up to 80 °C and briefly heat resistant up to 160 °C when surface is not mechanically stressed
These racks are 70 mm high.