In-line biocompatible guard cartridges
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-68242TI2 JR-68242TI
JOURJR-68242TI2EA 54.7 EUR
JOURJR-68242TI2 JOURJR-68242TI
In-line biocompatible guard cartridges
Kolommen voor chromatografie
The in-line PEEK guard cartridges extend the life of analytical columns by trapping contaminants from the sample and mobile phase, thereby preventing loss of retention. Only PEEK and titanium contacts the mobile phase and sample. This system is ideal for use with proteins or other biopolymers that may be contaminated or absorbed by steel surfaces.
  • Biocompatible
  • Easy to fill
These guard cartridges can be easily dry packed or slurry packed.