Bulb pipettes, MBL®, two marks, Class AS
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PRA620AS PRA622AS PRA624AS PRA626AS PRA630AS PRA632AS PRA634AS PRA636AS
BIBBPRA620ASEA 16.3 EUR
BIBBPRA620AS BIBBPRA622AS BIBBPRA624AS BIBBPRA626AS BIBBPRA630AS BIBBPRA632AS BIBBPRA634AS BIBBPRA636AS
Bulb pipettes, MBL®, two marks, Class AS
Pipetten Volpipetten
Soda lime glass.
  • Calibrated to deliver (TD, Ex)
  • Permanent amber stain marking
  • With +5 s waiting time
  • Features a safety bulb at the top of the pipette tube
  • Printed with batch code for downloadable batch certificate
Certificaten: Comply with ISO 648 and DIN 12690