Safety blood lancets, VITREX® STERILANCE® Press II
Leverancier: Vitrex Medical A/S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
393213 394213 395213 396213 399213
233-0125EA 17.6 EUR
233-0125 233-0126 233-0127 233-0128 MODU399213
Safety blood lancets, VITREX® STERILANCE® Press II
Bloedlancetten
Safety lancets for blood glucose and other in vitro diagnostic tests.
  • Needle fully shielded before and after use
  • Pre-loaded pressure activated design
  • High quality, ultra sharp tri-bevel needle, high speed penetration
A variety of point configurations and penetration depths suit most capillary blood requirements.