YOUTILITY-flessen, bruin glas, met schroefdop, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
218862859 218863658 218864457 218865453
215-3509EA 122 EUR
215-3509 215-3517 215-3526 215-3527
YOUTILITY-flessen, bruin glas, met schroefdop, DURAN®
Flessen Glazen flessen
DURAN®, Borosilicaatglas 3.3, bruin.
  • Amber coloured to protect light-sensitive materials
  • Specially shaped grip zones on the bottles for safer and easier handling
  • Ergonomic GL 45 screw cap made from food grade polypropylene
  • Shortened threads on bottle and cap for faster opening
  • Dedicated accessories for easy and simple identification and personalisation
  • Glass Type I according to USP, EP and JP
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
De DURAN® YOUTILITY-fles staat voor een nieuwe generatie glazen laboratoriumflessenMore Product Information
Order Now
Specificaties
Environmentally Preferable