Flessen, smalle hals, vierkant, met schroefdop, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Azlon®
115-1789EA 347 EUR
115-1789 115-1790 115-1792 115-1791
Flessen, smalle hals, vierkant, met schroefdop, Azlon®
Flessen Kunststof flessen
Polycarbonaat, zeer transparant.
  • Leakproof
  • Suitable for repeated sterilisation by autoclaving at +121 °C
  • Practical square design for economic use of space
  • Heavy-duty and durable
Ideal for the preparation and storage of biological media, reagents and aqueous solutions. With linerless polypropylene cap.