Connection system GL 45
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4507/45D 4541/04 4542/02 4537/02 4537/04 4537/06 4538/02 4538/04 4539/02 4539/04 4539/06 4539/08 4539/10 4541/02
BIBB4507/45DEA 47.8 EUR
BIBB4507/45D BIBB4541/04 BIBB4542/02 BIBB4537/02 BIBB4537/04 BIBB4537/06 BIBB4538/02 BIBB4538/04 BIBB4539/02 BIBB4539/04 BIBB4539/06 BIBB4539/08 BIBB4539/10 BIBB4541/02
Connection system GL 45
Sluitingen Flesdoppen
PP screwcaps for Pyrex® GL 45 media-lab bottles facilitate the transfer of liquids within a closed and sterile system.
  • Temperature resistant to 140 °C
  • Membrane screw cap is ideal for aerobic microbiology and cell culture, with gas permeable
  • Dishwasher safe
Ideal for chemistry and biotechnology laboratories. Components are fully interchangeable and compatible with both two and three port screwcaps. Four different tubing diameters can be accommodated (Ext.Ø; 1,6, 3,0, 3,2 and 6,0 mm). The additional pressure compensation set that includes a membraneMore Product Information