Racks for PCR and microcentrifuge tubes, double-sided, SP Bel-Art
Leverancier: Bel-Art Products, a part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Scienceware®
119-0459EA 234 EUR
119-0459 119-0458 119-0457 119-0464 119-0463 119-0462 119-0461 119-0460
Racks for PCR and microcentrifuge tubes, double-sided, SP Bel-Art
Rekken Tube Racks
PP, double-sided, reversible and fluorescent colours.
  • Rack colour assists in sample organisation and quick visual identification
  • Alphanumeric index allows easy tube identification
  • Matt side panel accepts tape or direct labelling
  • Moulded in handles provide a secure grip for transport
  • Can be used for freezer storage down to –80 °C
One side accommodates 0,5 ml microcentrifuge tubes and the opposite side holds 1,5 to 2,0 ml microcentrifuge tubes.