Magnetic stirring bars, octagonal, rare earth, Spinbar®, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F371020003 F371020238
BELAF371020003EA 70.3 EUR
BELAF371020003 BELAF371020238
Magnetic stirring bars, octagonal, rare earth, Spinbar®, SP Bel-Art
Roerders Magnetische roerstaven
These stirring bars reduce the frequency of spinout in viscous solutions or when stirring at high speeds. The high magnetic field strength of samarium cobalt (Rare Earth) magnets can provide up to three times stronger coupling versus Alnico magnetic stirring bars. Stronger coupling with the drive magnet keeps the stirring bar centered and rotating effectively.
  • Octagonal shape with pivot ring creates strong turbulence with minimal chattering and friction
  • Bars are easy to differentiate as encapsulated in bright green PTFE
  • Autoclave safe
Certificaten: More Product Information