Desiccator cabinets, Lab Companion™, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F424004001 F424004131 F424004111 F424004021 F424004121 F424004011 F424004031 F424004101
467-0141EA 3380 EUR
467-0141 467-0146 467-0144 467-0142 BELAF424004121 467-0143 467-0145 BELAF424004101
Desiccator cabinets, Lab Companion™, SP Bel-Art
Exsiccatoren Exsiccatorkasten
These compact, high-quality vacuum desiccators with clear or amber (UV blocking) polycarbonate maintain a vacuum and stay gastight allowing reliable dry storage or experiments over extended periods.
  • Stackable and provide vacuum integrity resulting in lower maintenance
  • Able to maintain 29 inches Hg vacuum for over 72 hours at room temperature
  • Single piece moulded design with no leakage and exceptional durability
  • Viton® 3-way valve offers consistent and uniform vacuum draw, vacuum release or gas exchange
  • Easy to clean, chemical resistant and allow quick visual check of the desiccator contents
  • Removable perforated shelves accommodate various storage needs
The desiccator cabinets have an analogue vacuum gauge on the front door which allows the vacuum level to be checked at a glance. Amber tinted UV blocking models block 100% of UVA, B, and C light to minimise damage or discolouration of light-sensitive samples. The hinged door enables easy access, built in locking tabs secure the door when not under vacuum. It can be used with any vacuum with free air capacity of 1,5 cubic feet per minute (CFM) ft or greater.
Bestelinformatie: Additional shelves, built-in ports (which permit attachment of up to three additional valves) and desiccant tray are available as optional accessories on request.