Bottles, wide neck, square, with screw cap
Leverancier: RIXIUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1-1301-0100-32 1-1301-0250-45 1-1301-0500-54 1-1301-1000-54
215-4751EA 90.9 EUR
215-4751 215-4752 215-4753 215-4754
Bottles, wide neck, square, with screw cap
Flessen Glazen flessen
Soda-lime glass, amber, with blue screw caps.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Closures in various designs are available on request
DIN thread. Space saving in storage and transport. Stable, non-slip. Amber to protect light-sensitive materials.