Tubing, Bioprene
Leverancier: WATSON MARLOW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
903.0064.032 903.0127.032 903.0096.032 903.E032.K24 903.E064.K24 903.E080.K24 903.E096.K24 903.0005.016 903.0005.J16 903.0080.J16
WATS903.0064.032EA 0 EUR
WATS903.0064.032 WATS903.0127.032 WATS903.0096.032 228-0503 228-0505 228-0506 228-0507 WATS903.0005.016 WATS903.0005.J16 WATS903.0080.J16
Tubing, Bioprene
Slangen Peristaltische pompslangen
Bioprene offers exceptionally long life and resistance to a wide range of chemicals. It is a first choice for applications including metering tablet-coating materials, pH control and media feed in fermentation and bioreactor metering, as well as pharmaceutical handling, dispensing, metering, transfer and filtration.
  • UV opaque
  • Fully weldable
  • No odour
  • Resistant to oxidising agents
Bioprene peristaltic pump tubing is fully approved for food and pharmaceutical applications. Validated processes are supported. Like Marprene, It lasts at least 10 times as long as tubing in other materials. Long-life tubing means stoppages for a tube change are rare events, easily fitted in with general maintenance.
Temperature resistant from 5 to 80 °C
Hardness (Shore durometer A): 66