Erlenmeyer Flasks with screw cap
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0748-10-2000 0748-10-3000 0748-10-025045
NEUB0748-10-2000EA 183 EUR
NEUB0748-10-2000 NEUB0748-10-3000 NEUB0748-10-025045
Erlenmeyer Flasks with screw cap
Kolven Erlenmeyers
Borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area. Screw cap with PTFE coated seal.