HPLC columns, MetaCarb
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A5244 A5215 A5220 A5220GC A5215GC A5235 A5235GC A5238 A5200 A5201 A5205 A5206 A5221 A5244GC A5210 A5210GC A5210250X046 A5238GC A5241 A5245 A5225 A5200GC A5203 A5205GC A5209 A5211 A5213 A5214 A5216 A5218 A5241GC
VARIA5244EA 2510 EUR
VARIA5244 VARIA5215 549-0877 VARIA5220GC VARIA5215GC 549-0878 VARIA5235GC VARIA5238 VARIA5200 VARIA5201 VARIA5205 VARIA5206 VARIA5221 VARIA5244GC VARIA5210 VARIA5210GC VARIA5210250X046 VARIA5238GC VARIA5241 VARIA5245 VARIA5225 VARIA5200GC VARIA5203 VARIA5205GC VARIA5209 VARIA5211 VARIA5213 VARIA5214 VARIA5216 VARIA5218 VARIA5241GC
HPLC columns, MetaCarb
Kolommen voor chromatografie
The MetaCarb™ line of carbohydrate columns provides high resolution columns for applications requiring high performance carbohydrate and organic acid analysis. These columns contain sulfonated polystyrene resins in the Calcium (87C), Hydrogen (87H), and Lead (87P) forms, and provide a wide range of selectivities for carbohydrate and organic analysis. They are widely used in the food and beverage industries for analysis of sweeteners, corn and cane sugars, fruit juices, soft drinks, beer and dairy products.
  • Superior carbohydrate separations
  • Wide range of column selectivities
  • Higher