Absorbent for acid spills, Chemizorb® H+
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.03874.2000 1.03874.5000
1.03874.2000EA 134 EUR
1.03874.2000 1.03874.5000
Absorbent for acid spills, Chemizorb® H+
Absorptiematerialen en bescherming tegen morsen
Chemizorb H+ is a granulate mixture that consists of a mineral co-polymer absorbent, an alkaline neutraliser, and a pH indicator.
  • Cover the spill with the amount necessary to neutralise and absorb
  • pH indicator makes it possible to monitor the neutralisation process
  • 100 g of Chemizorb H+ required to absorb and neutralise 100 ml of acid
The neutralisation process results in the release of CO₂. When the acid has been neutralised, the colour of the indicator changes from red to orange/yellow.