Protein assay kits, RED 660™
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-676 786-673 786-603 786-604
786-676EA 437 EUR
786-676 786-673 786-603 786-604
Protein assay kits, RED 660™
Testen Proteïne-assays
RED 660™ Protein Assay is a single reagent colorimetric assay that outperforms many commercial colorimetric assays, including Bradford and improved Coomassie/Bradford assays. It offers greater linearity, colour stability, and compatibility with detergents, reducing agents and other interfering agents compared to the Coomassie assays. The single, ready to use reagent allows for rapid analysis of total protein concentration and generates highly reproducible results.
The Neutralizer™ is a unique chemical that sequesters ionic detergents, including SDS, allowing the solution to be compatible with the RED 660™ Protein Assay. The Neutralizer™ can also be used with Laemmli loading buffer. Each vial is suitable for 10 ml RED 660™ Protein Assay reagent.
Order Now
Specificaties
Environmentally Preferable