Handschoenvoeringen, Puri ESD Palm Fit
Leverancier: PURITECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PFG510ESD/XL PFG500ESD/XL PFG500ESD/S PFG500ESD/M PFG500ESD/L PFG500ESD/XXL
PURTPFG510ESD/XLEA 121 EUR
PURTPFG510ESD/XL PURTPFG500ESD/XL PURTPFG500ESD/S PURTPFG500ESD/M PURTPFG500ESD/L PURTPFG500ESD/XXL
Handschoenvoeringen, Puri ESD Palm Fit
Handschoenen
Hand-specific for great fit
Excellent dexterity
Anti-static, prevent damaging static discharge
Protect humans and products from static electricity
  • Hand-specific for great fit
  • Excellent dexterity
  • Anti-static, prevent damaging static discharge
  • Protect humans and products from static electricity
Hand-specific for great fit
Excellent dexterity
Anti-static, prevent damaging static discharge
Protect humans and products from static electricity
Dikte van de coating: 0,1 mm.