Non-animal sera protein blocking Agent, FISH-Blocker™
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-674 786-675
786-674EA 594 EUR
786-674 786-675
Non-animal sera protein blocking Agent, FISH-Blocker™
Buffers Buffers voor elektroforese Blocking-buffers
FISH-Blocker™ uses a fish protein as the primary blocking agent to eliminate cross reactivity. Using a non mammalian protein eliminates or minimises the interaction of antibodies raised in mammals. It is offered either in TBS (Tris-buffered saline) or in PBS (phosphate-buffered saline) at pH 7,5.
  • Ready to use
  • Eliminates cross reactivity
  • Minimises the risk of non-specific binding