Disposable half masks against gas, vapours and particulates, FFA1/FFA2
Leverancier: Moldex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5120 5230 5430 5330
111-1707EA 39.7 EUR
111-1707 111-1708 111-1710 111-1709
Disposable half masks against gas, vapours and particulates, FFA1/FFA2
Ademhalingsmaskers Wegwerpmaskers Gasmaskers en gas- en stofmaskers
Maintenance-free compact half mask to protect against gas, vapour and dust.
  • Pre-assembled and ready for use
  • Lightweight dual-material construction
  • Adjusts to different face shapes automatically
  • Low profile design offers improved field of vision
Certificaten: EN 405:2001