Dispenser Bottles
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3804 3807
215-0243EA 47.4 EUR
215-0243 215-0284
Dispenser Bottles
Flesdispenseropzet
Bottles, LDPE, for quick dispensing, with PMP measuring cup. The 25 /50 ml measuring cup slides up and down the tube. The height of the tube determines the volume of liquid dispensed into the measuring cup when the bottle is squeezed.
  • Repetitive dispensing of the same volume of liquid
  • When the bottle pressure is released only the desired volume remains
  • Colour: Transparent /Red