Wafer tweezers, lockable
Leverancier: WINDRUSH TECHNOLOGY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M100-150L M100-200L M100-300L
FLUMM100-150LEA 0 EUR
FLUMM100-150L FLUMM100-200L FLUMM100-300L
Wafer tweezers, lockable
Pincetten
Manufactured entirely of PEEK, this unique design handles a delicate and fragile semiconductor wafer softly but firmly and without excessive contact. The contact area is optically-polished to reduce surface particle counts.
  • Suitable for continuous use up to 130 °C
  • No adhesive or metal parts
  • Lockable handle effectively reduces the forces imparted to the wafer