ESD safety shoes, Velcro®, Uvex1, 8542
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8542835 8542838 8542837 8542841 8542836 8542839 8542840 8542844 8542845 8542842 8542843 8542846 8542847 8542848 8542849 8542850 8542945 8542946 8542940 8542941 8542942 8542949 8542950 8542943 8542944 8542935 8542936 8542937 8542938 8542939 8542947 8542948 8542.8/51 8542.8/52
UVEX8542835EA 116 EUR
UVEX8542835 UVEX8542838 UVEX8542837 UVEX8542841 UVEX8542836 UVEX8542839 UVEX8542840 UVEX8542844 UVEX8542845 UVEX8542842 UVEX8542843 UVEX8542846 UVEX8542847 UVEX8542848 UVEX8542849 UVEX8542850 UVEX8542945 UVEX8542946 UVEX8542940 UVEX8542941 UVEX8542942 UVEX8542949 UVEX8542950 UVEX8542943 UVEX8542944 UVEX8542935 UVEX8542936 UVEX8542937 UVEX8542938 UVEX8542939 UVEX8542947 UVEX8542948 UVEX8542.8/51 UVEX8542.8/52
ESD safety shoes, Velcro®, Uvex1, 8542
Schoenen en laarzen
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, and a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black
Width: 11 or 12
Certificaten: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1, SRC