Blocking solution, BLOK™ Casein
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-194 786-196
786-194EA 363 EUR
786-194 786-196
Blocking solution, BLOK™ Casein
Buffers Buffers voor elektroforese Blocking-buffers
BLOK™ Casein is a purified protein, used for blocking free binding sites in Western blot, ELISA plate well, immunohistochemistry and other immunochemical applications.
  • Ready to use
  • Available in the choices of TBS or PBS buffers