Electrophoresis gel stain, Reversible Zinc Stain™
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-32DSZN 786-32ZN
786-32DSZNEA 459 EUR
786-32DSZN 786-32ZN
Electrophoresis gel stain, Reversible Zinc Stain™
Kleurstoffen Kleuringen voor elektroforese
A single step stain for the rapid detection of proteins resolved by PAGE. The stain is based on the interaction of zinc ions with polyacrylamide and proteins.
  • Reversible, simple, fast, and sensitive
  • High contrast protein bands
  • For proteins difficult to detect with silver stains such as glycoproteins
This stain works with native, SDS denatured gels, and gels containing glycine, tricine and primary amine buffers.