ESD safety ankle boots, lace-up, uvex1, 8545
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8545.8/51 8545.8/40 8545.8/35 8545.8/36 8545.8/37 8545.8/39 8545.8/52 8545.8/41 8545.8/42 8545.8/43 8545.8/44 8545.8/45 8545.8/46 8545.8/47 8545.8/48 8545.8/49 8545.8/50 8545.8/38
UVEX8545.8/51EA 125 EUR
UVEX8545.8/51 UVEX8545.8/40 UVEX8545.8/35 UVEX8545.8/36 UVEX8545.8/37 UVEX8545.8/39 UVEX8545.8/52 UVEX8545.8/41 UVEX8545.8/42 UVEX8545.8/43 UVEX8545.8/44 UVEX8545.8/45 UVEX8545.8/46 UVEX8545.8/47 UVEX8545.8/48 UVEX8545.8/49 UVEX8545.8/50 UVEX8545.8/38
ESD safety ankle boots, lace-up, uvex1, 8545
Schoenen en laarzen
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, and a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • Elastic laces for quick and individual adjustment
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black
Width: 11
Certificaten: EN 61340 (ESD); DGUV 112-191; EN 20345:2011; S2, SRC