Sieve Standards, Polydisperse Particle Standard
Leverancier: WHITEHOUSE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PS194 PS237 PS181 PS205 PS227 PS225 PS180 PS315 PS202 PS313 PS316 PS314
WHITPS194EA 861 EUR
WHITPS194 WHITPS237 WHITPS181 WHITPS205 WHITPS227 WHITPS225 WHITPS180 WHITPS315 WHITPS202 WHITPS313 WHITPS316 WHITPS314
Sieve Standards, Polydisperse Particle Standard
Standaarden Standaarden voor particelgrootte
The Polydisperse particle standards formally known as the BCR 'Mirror' standards were made both to assess the performance of the analytical laboratories and to augment a limited supply of the official standards. Although sub-divided, the BCR samples were never certified, which leaves these new polydisperse particle standards as the most comprehensively analysed set of spherical particle size reference standards available.
  • Calibration for any method of particle size analysis
  • Unambiguous primary methods used - microscopy, sieving, sedimentation and the Coulter method
Traceability through NIST, BCR and NPL standards.