Boiling baths, Precisterm
Leverancier: SELECTA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6000141 6000388 6000386
SELE6000141EA 1130 EUR
SELE6000141 SELE6000388 SELE6000386
Boiling baths, Precisterm
Baden Kookbaden
Double skinned AISI 310 stainless steel tank with an exterior case of AISI 304 stainless steel. The heating elements embedded in INCOLOY heat and corrosion resistant stainless steel.
  • Illuminated mains power switch
  • Hydraulic thermostat temperature control, synchronised with the 'ON' indicator lamp
  • Safety thermostat cut out indicator lamp (DIN 12877.2)
Order Now
Specificaties
Environmentally Preferable