Gel drying solution for protein gels
Leverancier: G-Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-685 786-686
786-685EA 264 EUR
786-685 786-686
Gel drying solution for protein gels
Gel voor elektroforese Voorgegoten gels
This gel drying solution effectively and evenly dries protein electrophoresis gels by the mechanism of passive evaporation and is suitable for permanent gel storage and densitometry.
  • Ready to use
  • Regulates rate of drying
  • Prevents cracking of the gel
  • Compatible with all drying membranes