Wash buffers, femto TBST™ and femto PBST™
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-161 786-162
786-161EA 117 EUR
786-161 786-162
Wash buffers, femto TBST™ and femto PBST™
Buffers Buffers voor elektroforese Wasbuffers
The femto TBST™ and femto PBST™ wash buffers will enhance sensitivity of immunoassays by minimising the 'washing out' of immuno agents or immuno complexes, a common problem associated with classical TBST and PBST buffers. The wash buffers aid in the generation of cleaner backgrounds, resulting in a larger ratio of signal to background. These can be used for incubation and washing procedures.