Specialty trolleys
Leverancier: TRESTON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C74007004 C74041003 C74007102 C74007105
SOVEC74007004EA 0 EUR
SOVEC74007004 SOVEC74041003 SOVEC74007102 SOVEC74007105
Specialty trolleys
Transportwagens
The specialty trolleys can be equipped with shelves, tops and other accessories to fit any required application such as parts transportation or tool storage.
  • Four swivel castors (two with brakes) of Ø 150 mm
  • Load capacity: 349 kg