One-piece hex-head fittings
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-55100-5 JR-55110-5
JOURJR-55100-5EA 90.2 EUR
JOURJR-55100-5 JOURJR-55110-5
One-piece hex-head fittings
Fittingen en connectoren
One-piece hex-head fittings made from PEEK are easy to use with the ValvTool and the Color-It Color-Coded Fingertight adapters. The 10-32 thread is suitable for standard high-pressure female connections. They are available both in a short and long version.
  • Ideal for use with PEEK tubing
  • Withstand pressures up to 350 bar (5000 psi)
  • For any high pressure fittings