Dissolved oxygen probes, InPro® 6800
Leverancier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
52200968 52200966
METN52200968EA 0 EUR
METN52200968 METN52200966
Dissolved oxygen probes, InPro® 6800
Elektroden Elektroden voor opgeloste zuurstof
The InPro O2 sensor family provides the means for reliable inline measurement of dissolved oxygen in a wide spectrum of processes that operate under sterile/hygienic conditions.
  • Hygienic design, capable of withstanding CIP procedures
  • Stainless steel surfaces with highly polished finish
  • Steam sterilisable and autoclavable up to 140°C
  • Low detection limit at high signal stability
  • O-rings with FDA and USP Class VI approval
  • Minimal maintenance requirement
  • ISM (Intelligent Sensor Management)