Wrenches, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A-304 A-305 A-320
UPCHA-304EA 35.2 EUR
UPCHA-304 UPCHA-305 UPCHA-320
Wrenches, Upchurch Scientific®
Moersleutels
These standard size wrenches can be used to tighten most nuts into unions.